Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NSC 405020
GREENER_CHOICESDP

Catalog No. 490210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-0210 10 mg
49-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-0210 Supplier Tocris Bioscience™ Supplier No. 4902/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Histone deacetylase inhibitor

Membrane type-1 matrix metalloproteinase (MT1-MMP) inhibitor (IC50 > 100 μmol/L). Directly interacts with the hemopexin domain (PEX) of MT1-MMP, affecting homodimerization and repressing its pro-tumorigenic activity in vivo. Displays no effect on the catalytic activity of MT1-MMP or MMP-2.

Specifications

Chemical Name or Material NSC 405020
CAS 7497-07-6
Synonym 2-methyl-5-chlorobenzoic acid, 5-chloro-o-toluic acid, 5-chloro-2-methyl-benzoic acid, benzoic acid, 5-chloro-2-methyl, o-toluic acid, 5-chloro, 5-chloro-2-methylbenzoicacid, 5-chloro-2-methyl benzoic acid, 3-chloro-6-methylbenzoic acid, pubchem4578, ksc173q5r
Quantity 10 mg
Molecular Formula C12H15Cl2NO
Formula Weight 260.16
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.