Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NSC 636819
GREENER_CHOICESDP

Catalog No. 528750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
528750 50 mg
528710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 528750 Supplier Tocris Bioscience™ Supplier No. 5287/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

KDM4A/KDM4B inhibitor

NSC 636819 is a KDM4A/KDM4B inhibitor; inhibits histone demethylase activity on H3K9me3 and induces apoptosis in LNCaP prostate cancer cells in vitro. Also negatively regulates androgen-responsive genes.

Specifications

Chemical Name or Material 1,5-Bis[(1E)-2-(3,4-dichlorophenyl)ethenyl]-2,4-dinitrobenzene
CAS 1618672-71-1
Quantity 50 mg
Target Histone Demethylase Inhibitors
Molecular Formula C22H12Cl4N2O4
Formula Weight 510.15
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.