Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NSC 663284
SDP

Catalog No. 186710 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-671-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-671-0 Supplier R&D Systems™ Supplier No. 1867/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective Cdc25 phosphatase inhibitor

Potent, selective inhibitor of Cdc25 dual specificity phosphatases (Ki values are 29, 95 and 89 nM for human Cdc25A, Cdc25B2 and Cdc25C respectively); > 20- and > 450-fold selective over VHR and PTP1B phosphatases respectively. Arrests cells at both G1 and G2/M phase and blocks cdk2 and cdk1 activation. Blocks proliferation of a range of human tumor cell lines (IC50 = 0.2 - 35 μM).

Chemical Identifiers

CAS 383907-43-5
Molecular Formula C15H16ClN3O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 321.76
Purity >99%
Molecular Weight (g/mol) 321.76
Chemical Name or Material NSC 663284
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.