Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NSC 668036
GREENER_CHOICE

Catalog No. 581310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-131-0 10 mg
58-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-131-0 Supplier Tocris Bioscience™ Supplier No. 5813/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein

NSC 668036 is a Wnt signaling inhibitor. Binds the PDZ domain of the Dishevelled (Dvl) protein. Inhibits proliferation, migration and TGF-β-induced differentiation of fibroblasts in vitro. Reduces severity of pulmonary fibrosis in a mouse model.

Specifications

Chemical Name or Material N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanine-(1S)-1-carboxy-2-methylpropyl ester
CAS 144678-63-7
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C21H36N2O9
Formula Weight 460.52
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.