Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
NSC 668036 is a Wnt signaling inhibitor. Binds the PDZ domain of the Dishevelled (Dvl) protein. Inhibits proliferation, migration and TGF-β-induced differentiation of fibroblasts in vitro. Reduces severity of pulmonary fibrosis in a mouse model.
Specifications
Specifications
| Chemical Name or Material | N-[(1,1-dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanine-(1S)-1-carboxy-2-methylpropyl ester |
| CAS | 144678-63-7 |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C21H36N2O9 |
| Formula Weight | 460.52 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

