Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540725 Supplier Invitrogen™ Supplier No. PA540725
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KYNVGDLVWS KVSGYPWWPC MVSADPLLHS YTKLKGQKKS ARQYHVQFFG Sequence homology: Dog: 93%; Guinea Pig: 88%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 76%; Rat: 86%.

This gene encodes a protein that contains four domains present in other developmental proteins: a PWWP domain, an HMG box, a SET domain, and a PHD-type zinc finger. It is expressed ubiquitously in early development. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene maps to the 165 kb WHS critical region and has also been involved in the chromosomal translocation t(4;14)(p16.3;q32.3) in multiple myelomas. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. Some transcript variants are nonsense-mediated mRNA (NMD) decay candidates, hence not represented as reference sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NSD2
Gene Accession No. O96028
Gene Alias 5830445G22Rik; 9430010A17Rik; AW555663; C130020C13Rik; D030027O06Rik; D930023B08Rik; Histone-lysine N-methyltransferase NSD2; histone-lysine N-methyltransferase NSD2; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; Kiaa1090; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; mKIAA1090; MMSET; multiple myeloma SET domain containing protein type III; multiple myeloma SET domain-containing protein; NSD2; nuclear receptor binding SET domain protein 2; nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; putative histone-lysine N-methyltransferase NSD2-like protein; REIIBP; RGD1565590; trithorax/ash1-related protein 5; TRX5; WHS; Whsc1; Whsc1l; Wolf-Hirschhorn syndrome candidate 1; Wolf-Hirschhorn syndrome candidate 1 (human); Wolf-Hirschhorn syndrome candidate 1 protein; Wolf-Hirschhorn syndrome candidate 1 protein homolog
Gene Symbols NSD2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human WHSC1 (aa 219-268).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7468
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.