Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSD3 Antibody (7B4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104324AF350
Description
NSD3 Monoclonal specifically detects NSD3 in Human samples. It is validated for Western Blot, ELISA.Specifications
NSD3 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
WHSC1L1 | |
Recombinant human NSD3 (383-660aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
IgG2b κ |
Western Blot, ELISA | |
7B4 | |
Western Blot, ELISA | |
EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1 | |
Mouse | |
Protein G purified | |
Cancer | |
54904.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction