Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSD3 Antibody (7B4), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104324C
Description
NSD3 Monoclonal specifically detects NSD3 in Human samples. It is validated for Western Blot, ELISA.Specifications
NSD3 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
WHSC1L1 | |
Recombinant human NSD3 (383-660aa) purified from E. coli | |
0.1 mL | |
Cancer | |
54904 | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
7B4 | |
Western Blot, ELISA | |
EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human NSD3 mAb, clone 2E9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NSD3 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction