Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582643 Supplier Invitrogen™ Supplier No. PA582643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTQPEQTNAG EVASSLSSTE IRRHSQRRHT SAEEEEPPPV KIAWKTAAAR KSLPASITMH KGSLDLQKCN MSPVVKIEQV FALQNATGDG KFIDQFVYST KGIGNKTEIS VRGQDRLIIS TPNQRNEKPT QSVSSPEATS Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 92%.

This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSD3
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nsd3
Gene Accession No. Q9BZ95
Gene Alias 6720429E03; A530023P05; AI528490; DC28; DKFZp667H044; FLJ20353; Histone-lysine N-methyltransferase NSD3; MGC126766; MGC142029; NSD3; nuclear receptor binding SET domain protein 3; nuclear SET domain-containing protein 3; pp14328; Protein whistle; WHISTLE; WHSC1L1; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like 1; Wolf-Hirschhorn syndrome candidate 1-like 1 (human); Wolf-Hirschhorn syndrome candidate 1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1 homolog
Gene Symbols Nsd3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NSD3. Recombinant protein control fragment (Product #RP-101026).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.