Learn More
Invitrogen™ NSD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117022
Description
Positive Samples: 293T, HepG2, HeLa, MCF7, Mouse liver, Mouse brain, Mouse spleen, Mouse kidney, Mouse lung Immunogen sequence: EEALSQAKKS VASKTEVKKT RRPRSVLNTQ PEQTNAGEVA SSLSSTEIRR HSQRRHTSAE EEEPPPVKIA WKTAAARKSL PASITMHKGS LDLQKCNMSP VVKIEQVFAL QNATGDGKFI DQFVYSTKGI GNKTEISVRG QDRLIISTPN QRNEKPTQSV SSPEATSGST GSVEKKQQRR SIRTRSESEK STEVVPKKKI K.
This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.
Specifications
NSD3 | |
Polyclonal | |
Unconjugated | |
Nsd3 | |
6720429E03; A530023P05; AI528490; DC28; DKFZp667H044; FLJ20353; Histone-lysine N-methyltransferase NSD3; MGC126766; MGC142029; NSD3; nuclear receptor binding SET domain protein 3; nuclear SET domain-containing protein 3; pp14328; Protein whistle; WHISTLE; WHSC1L1; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like 1; Wolf-Hirschhorn syndrome candidate 1-like 1 (human); Wolf-Hirschhorn syndrome candidate 1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
234135, 290831, 54904 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.11 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q6P2L6, Q9BZ95 | |
Nsd3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 400-600 of human NSD3/NSD3/WHSC1L1 (NP_0602482). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.