Learn More
Invitrogen™ NSE Polyclonal Antibody

Description
Immunogen sequence: VIKDKYGKDA TNVGDEGGFA PNILENSEAL ELVKEAIDKA GYTEKIVIGM DVAASEFYRD GKYDLDFKSP TDPSRYITGD QLGALYQDFV RDYPVVSIED PFDQDDWAAW SKFTANVGIQ IVGDDLTVTN PKRIERAVEE KACNCLLLKV NQIGSVTEAI QACKLAQENG WGVMVSHRSG ETEDTFIADL VVGLCTGQIK TGAPCRSERL AKYNQLMRIE EELGDEARFA GHNFRNPSVL; Positive Samples: HeLa, LO2, SH-SY5Y, U-251MG, Mouse skeletal muscle, Mouse liver, Mouse brain, Rat brain, Rat skeletal muscle; Cellular Location: Cell membrane, Cytoplasm.
Specifications
Specifications
| Antigen | NSE |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | ENO2 |
| Gene Accession No. | P07323, P09104, P17183 |
| Gene Alias | 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; AI837106; D6Ertd375e; Eno; ENO2; Eno-2; ENOG; Enolase 2; enolase 2 (gamma, neuronal); enolase 2, gamma neuronal; enolase 2, gamma, neuronal; epididymis secretory protein Li 279; gamma enolase; gamma-enolase; gamma-enolase-like; gamma-subunit of enolase; HEL-S-279; LOC100911625; Neural enolase; neuron specific enolase; neuron specific gamma enolase; Neuronal Enolase; neuronal enriched enolase; neurone-specific enolase; neuron-specific enolase; NSE; RNEN3 |
| Gene Symbols | ENO2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.