Learn More
Invitrogen™ NSE2 Monoclonal Antibody (OTI4A2)

Description
NSE2 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | NSE2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4A2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Nsmce2 |
| Gene Accession No. | Q96MF7 |
| Gene Alias | 1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.