Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544399 Supplier Invitrogen™ Supplier No. PA544399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KFLALQSKNS DADFQNNEKF VQFKQQLKEL KKQCGLQADR EADGTEGVDE Sequence homology: Cow: 86%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 100%.

This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSE2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nsmce2
Gene Accession No. Q96MF7
Gene Alias 1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7
Gene Symbols Nsmce2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human NSMCE2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286053
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.