Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579765
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579765 Supplier Invitrogen™ Supplier No. PA579765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, human HepG2 cell. IHC: mouse testis tissue, rat testis tissue, human glioma tissue, human intestinal cancer tissue, human mammary cancer tissue.

Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSF
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene NSF
Gene Accession No. P46459, P46460, Q9QUL6
Gene Alias AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF
Gene Symbols NSF
Host Species Rabbit
Immunogen E. coli-derived human NSF recombinant protein (Position: N620-D744).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18195, 4905, 60355
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.