Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595645
Description
Antigen Affinity Purified from Pooled Serum.
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
Specifications
NSF | |
Polyclonal | |
Unconjugated | |
NSF | |
AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60355 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.25 mg/mL | |
10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q9QUL6 | |
NSF | |
Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction