Learn More
Invitrogen™ NSF Polyclonal Antibody

Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | NSF |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 10 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | NSF |
| Gene Accession No. | P18708, P46459, P46460, Q9QUL6 |
| Gene Alias | AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF |
| Gene Symbols | NSF |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.