Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576126
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
Specifications
NSF | |
Polyclonal | |
Unconjugated | |
NSF | |
AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18195, 4905, 60355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P46459, P46460, Q9QUL6 | |
NSF | |
Recombinant protein of human NSF. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction