Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981211
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1211 Supplier Bioss Supplier No. BS11255R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.

Specifications

Antigen NSF
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NSF
Gene Accession No. P46459
Gene Alias AI316878; AU020090; AU067812; BOS_19314; Erg1; LOC100353806; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; Suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; Vesicular-fusion protein NSF
Gene Symbols NSF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NSF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4905
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.