Learn More
Invitrogen™ NSFL1C Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (83%), Pig (98%), Chicken (87%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | NSFL1C |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Nsfl1c |
| Gene Accession No. | O35987, Q9CZ44, Q9UNZ2 |
| Gene Alias | dJ776F14.1; Munc-18c; NSFL1 (p97) cofactor (p47); NSFL1 cofactor; NSFL1 cofactor p47; NSFL1C; p47; p97 cofactor p47; RP4-776F14.2; SHP1 homolog; Stxbp3a; UBX domain-containing protein 2C; UBX1; UBXD10; UBXN2C; XY body-associated protein XY40; XY40 |
| Gene Symbols | Nsfl1c |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.