Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536497 Supplier Invitrogen™ Supplier No. PA536497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 21 kDa. Purity is >95% by SDS-PAGE.

NEEP21 (neuron-enriched endosomal 21kDa protein), also known as brain neuron cytoplasmic protein 1, NSG1 (neuron-specific proteins family member 1), P21 or D4S234E, is a single pass type II membrane protein belonging to the NSG family. It is highly expressed during neuronal maturation but its expression is downregulated in adult tissues. NEEP21 predominantly localizes to Rab 4-positive early endosomes in the somatodendritic neuronal compartment and is essential for proper receptor sorting and recycling in neurons. It associates with GRIP1 and GluR-2 and mediates the surface expression of GluR-2. When this interaction is interrupted, GluR-2 accumulates in early endosomes and leads to changes in evoked synaptic current properties. In addition, NEEP21 forms a complex with the SNARE protein, Syntaxin 13 (also known as Syntaxin 12), and participates in the recycling of transferrin receptors (TFRs) and NTR2 (Neurotensin receptor 2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NSG1
Gene Accession No. P02683, P42857, Q62092
Gene Alias brain neuron cytoplasmic protein 1; brain neuron cytoplasmic protein 1/2; brain specific mRNA b; Bsmrb; carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein; D4S234; D4S234E; m234; Neep21; neuron specific gene family member 1; Neuronal vesicle trafficking-associated protein 1; Neuron-enriched endosomal protein of 21 kDa; Neuron-specific protein family member 1; NSG1; p21; PEP1
Gene Symbols NSG1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 130-180 of human NEEP21.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18196, 25247, 27065
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.