Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583825 Supplier Invitrogen™ Supplier No. PA583825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAFTYDHSCP EGFVYKHKRC IPASLDAYYS SQDPNSRSRF YTVISHYSVA KQSTARAIGP WLSAAAVIHE PKPPKTQGH Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

NSG2 is a protein coding gene. Gene ontology (GO) annotation include cytoplasmic vesicle membrane; dendrite; early endosome; early endosome membrane; endosome; Golgi apparatus; Golgi cis cisterna membrane; integral component of membrane; late endosome; lysosomal lumen; multivesicular body membrane; trans-Golgi network membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSG2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NSG2
Gene Accession No. Q9Y328
Gene Alias CALY3; HMP19; HMP19 protein; hypothalamus golgi apparatus expressed 19 kDa protein; neuron specific gene family member 2; Neuronal vesicle trafficking-associated protein 2; neuron-specific protein family member 2; NSG2; p19 protein; protein 8.5; Protein p19
Gene Symbols NSG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NSG2. Recombinant protein control fragment (Product #RP-108374).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.