Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981890
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1890 Supplier Bioss Supplier No. BS12036R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NSG2 is a protein coding gene. Gene ontology (GO) annotation include cytoplasmic vesicle membrane; dendrite; early endosome; early endosome membrane; endosome; Golgi apparatus; Golgi cis cisterna membrane; integral component of membrane; late endosome; lysosomal lumen; multivesicular body membrane; trans-Golgi network membrane.

Specifications

Antigen NSG2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NSG2
Gene Accession No. Q9Y328
Gene Alias CALY3; Hmp19; HMP19 protein; hypothalamus golgi apparatus expressed 19 kDa protein; neuron specific gene family member 2; Neuronal vesicle trafficking-associated protein 2; Neuron-specific protein family member 2; NSG2; p19 protein; Protein 8.5; Protein p19
Gene Symbols NSG2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NSG2/HMP19.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51617
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.