Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nSMase Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP297998AF405

Catalog No. NB050728


Only null left
Add to Cart

Description

Description

nSMase Polyclonal specifically detects nSMase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

nSMase
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin
EC 3.1.4.12, ISC1, lyso-PAF-PLC, Lyso-platelet-activating factor-phospholipase C, Neutral sphingomyelinase, N-SMase, NSMASE, NSMASE1, sphingomyelin phosphodiesterase 2, sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)
Produced in rabbits immunized with E. coli-derived Human nSMase fragment.
0.1 mL
Lipid and Metabolism
6610
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 405
50mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.