Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSMCE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560234
Description
Immunogen sequence: EQYIRETYPD AVKICNICHS LLIQGQSCET CGIRMHLPCV AKYFQSNAEP RCPHCNDYWP HEIPKVFDPE KERESGVLKS NK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 87%.

Specifications
NSMCE1 | |
Polyclonal | |
Unconjugated | |
NSMCE1 | |
2510027N19Rik; HSPC333; HSPC337; Non-SMC element 1 homolog; non-SMC element 1 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 1 homolog; NSE1; NSE1 homolog, SMC5-SMC6 complex component; Nsmce1; RGD1307760 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
197370 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WV22 | |
NSMCE1 | |
Recombinant protein corresponding to Human NSMCE1. Recombinant protein control fragment (Product #RP-98762). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction