Learn More
Invitrogen™ NSMCE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590865
Description
Immunogen sequence: RMGVMTDVHR RFLQLLMTHG VLEEWDVKRL QTHCYKVHDR NATVDKLEDF INNINSVLES LYIEIKRGVT EDDGRPIYAL VNLATTSISK MATDFAENEL DLFRKALELI IDSETGFASS TNILNLVDQL KGKKMRKKEA EQVLQKFVQN KWLIEKEGEF TLHGRAILEM EQYIRETYPD AVKICNICHS LLIQGQSCET CGIRMHLPCV AKYFQSNAEP RCPHCNDYWP HEIPKVFDPE KERESGVLKS NKKSLRSRQH; Positive Samples: BxPC-3, Rat testis; Cellular Location: Chromosome, Nucleus, telomere.
The NSMCE1 (Non-SMC Element 1 Homolog) gene encodes a protein that is a key component of the SMC5-SMC6 complex, which is essential for DNA repair and the maintenance of genomic stability. Located on chromosome 16, the NSMCE1 protein interacts with other components of the SMC5-SMC6 complex, including SMC5, SMC6, and NSMCE2, facilitating the repair of DNA double-strand breaks and promoting chromosome segregation during cell division. NSMCE1 is particularly involved in homologous recombination, a critical pathway for error-free DNA repair. Mutations or deficiencies in NSMCE1 can lead to genomic instability, increased sensitivity to DNA-damaging agents, and have been associated with several genetic disorders and cancer predispositions.
Specifications
NSMCE1 | |
Polyclonal | |
Unconjugated | |
NSMCE1 | |
2510027N19Rik; HSPC333; HSPC337; Non-SMC element 1 homolog; non-SMC element 1 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 1 homolog; NSE1; NSE1 homolog, SMC5-SMC6 complex component; Nsmce1; RGD1307760 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
197370, 361645 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
4.03 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q499U6, Q8WV22 | |
NSMCE1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 7-266 of human NSMCE1 (NP_6595472). | |
100 μL | |
Primary | |
Human, Rat, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.