Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSMCE4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557954
Description
Immunogen sequence: FLLGSIYGEC PVPKPRVDRP RKVPVIQEER AMPAQLRRME ESHQEATEKE VERILGLLQT YFREDPDTPM SFFDFVVDPH SFP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
NSMCE4A (NSE4 Homolog A, SMC5-SMC6 Complex Component) is a Protein Coding gene. Diseases associated with NSMCE4A includeChromosome 10Q26 Deletion Syndrome. Among its related pathways areMetabolism of proteinsandSUMOylation.
Specifications
NSMCE4A | |
Polyclonal | |
Unconjugated | |
NSMCE4A | |
2410003A14Rik; C10orf86; CH73-19J14.1; hypothetical protein LOC527312; hypothetical protein LOC553544; LOW QUALITY PROTEIN: non-structural maintenance of chromosomes element 4 homolog A; non-SMC element 4 homolog A; non-SMC element 4 homolog A (S. cerevisiae); non-structural maintenance of chromosomes element 4 homolog A; NS4EA; NSE4 homolog A; NSE4 homolog A, SMC5-SMC6 complex component; NSE4 homolog, SMC5-SMC6 complex component A; NSE4A; NSMCE4A; PP4762; RGD1309025; zgc:109897 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54780 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXX6 | |
NSMCE4A | |
Recombinant protein corresponding to Human NSMCE4A. Recombinant protein control fragment (Product #RP-97035). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction