Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSMCE4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111097 Supplier Invitrogen™ Supplier No. PA5111097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FHVSFIIRDG FARIRLDQDR LPVIEPVSIN EENEGFEHNT QVRNQGIIAL SYRDWEEIVK TFEISEPVIT PSQRQQKPSA Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 83%.

NSMCE4A (NSE4 Homolog A, SMC5-SMC6 Complex Component) is a Protein Coding gene. Diseases associated with NSMCE4A includeChromosome 10Q26 Deletion Syndrome. Among its related pathways areMetabolism of proteinsandSUMOylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSMCE4A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NSMCE4A
Gene Accession No. Q9NXX6
Gene Alias 2410003A14Rik; C10orf86; CH73-19J14.1; hypothetical protein LOC527312; hypothetical protein LOC553544; LOW QUALITY PROTEIN: non-structural maintenance of chromosomes element 4 homolog A; non-SMC element 4 homolog A; non-SMC element 4 homolog A (S. cerevisiae); non-structural maintenance of chromosomes element 4 homolog A; NS4EA; NSE4 homolog A; NSE4 homolog A, SMC5-SMC6 complex component; NSE4 homolog, SMC5-SMC6 complex component A; NSE4A; NSMCE4A; PP4762; RGD1309025; zgc:109897
Gene Symbols NSMCE4A
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human NSMCE4A. Recombinant protein control fragment (Product #RP-105950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.