Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSP1 Monoclonal Antibody (32D6)
GREENER_CHOICE

Catalog No. 01670286
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-286 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-286 Supplier Invitrogen™ Supplier No. MA110029
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10029 detects Nsp1p from S. cerevisiae samples. To confirm a high affinity for Nsp1p, strain RS453, which expresses a shortened isoform of Nsp1p, was compared to the wildtype strain BJ5465. NSP1 in the RS453 strain contains an internal deletion that removes the coding sequence for 6 FXFG repeats, which are not essential for protein function, and encodes a protein that has been observed to migrate at ~85kDa on SDS-PAGE gels. Wildtype Nsp1p has a predicted MW of 86.5 kDa but has been observed to migrate on SDS gels at ~100 kDa. SDS-PAGE testing results showed wild type Nsp1p at ~108 kDa and Nsp1p RS453 at ~91 kDa, confirming specificity for the Nsp1p protein. For western blots of yeast protein samples, use MA1-10029 diluted 1:10,000 (cell lysates) to 1:50,000 (nuclear fractions), followed by chemiluminescent detection (ECL). For other (non-ECL) western detection methods, try MA1-10029 diluted 1:1,000 to 1:5,000. For immunofluorescence on yeast cells, use MA1-10029 diluted 1:1,000 to 1:10,000.

Nucleoporin NSP1 is a component of the yeast nuclear pore complex (NPC). NPC components, collectively referred to as nucleoporins (NUPs), can play the role of both NPC structural components and of docking or interaction partners for transiently associated nuclear transport factors. Active directional transport is assured by both, a Phe-Gly (FG) repeat affinity gradient for these transport factors across the NPC and a transport cofactor concentration gradient across the nuclear envelope (GSP1 and GSP2 GTPases associated predominantly with GTP in the nucleus, with GDP in the cytoplasm). NSP1 plays an important role in several nuclear transport pathways including poly(A)+ RNA, tRNA, pre-ribosome, signal recognition particle (SRP), and protein transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSP1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 32D6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 5mM sodium azide
Gene NSP1
Gene Accession No. P14907
Gene Alias FG-nucleoporin NSP1; G7001; J1207; NSP1; NSR1; Nsr1p; Nuclear localization sequence-binding protein; Nuclear pore protein NSP1; Nucleoporin NSP1; Nucleoskeletal-like protein; p110; p67; SHE5; YGR159C; YJL041W
Gene Symbols NSP1
Host Species Mouse
Immunogen Yeast nuclear preparation.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 853409
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.