Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983054
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3054 Supplier Bioss Supplier No. BS13657R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Eph receptor-binding protein which may be a positive regulator of TCR signaling. Binding to BCAR1 is required to induce membrane ruffling and promote EGF-dependent cell migration (By similarity).

Specifications

Antigen NSP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SH2D3C
Gene Accession No. Q8N5H7
Gene Alias Cas/HEF1-associated signal transducer; Chat; Novel SH2-containing protein 3; NSP3; PRO34088; SH2 domain containing 3C; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; Sh2d3c; Shep1; UNQ272/PRO309/PRO34088
Gene Symbols SH2D3C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NSP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10044
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.