Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSP5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582761
Description
Immunogen sequence: APREFSVTVS RERSVPRGPS NPRKSVSSPT SSNTPTPTKH LRTPSTKPKQ ENEGGEKAAL ESQVRELLAE AKAKDSEINR LRSELKKYKE KRTLNA.
The protein encoded by this gene belongs to the cytospin-A family. It is localized in the nucleus, and highly expressed in testis and some cancer cell lines. A chromosomal translocation involving this gene and platelet-derived growth factor receptor, beta gene (PDGFRB) may be a cause of juvenile myelomonocytic leukemia. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
NSP5 | |
Polyclonal | |
Unconjugated | |
Specc1 | |
2810012G08Rik; B230396K10Rik; cytokinesis and spindle organization B; cytospin B; cytospin-B; CYTSB; HCMOGT1; HCMOGT-1; NSP; NSP5; NSP5a3a; NSP5alpha3alpha; Nuclear structure protein 5; RGD1309718; SPECC1; spectrin domain with coiled-coils 1; sperm antigen HCMOGT-1; Sperm antigen with calponin homology and coiled-coil domains 1; sperm antigen with calponin-like and coiled coil domains 1; structure protein NSP5a3a; structure protein NSP5a3b; structure protein NSP5b3a; structure protein NSP5b3b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92521 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5M775 | |
Specc1 | |
Recombinant protein corresponding to Human SPECC1. Recombinant protein control fragment (Product #RP-93234). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction