Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
nSR100 (SRRM4), Rabbit anti-Mouse, Human, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN147725UG
Description
Anti-nSR100 (SRRM4), Cat. No. ABN1477, is a highly specific rabbit polyclonal antibody that targets Serine/arginine repetitive matrix protein 4 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Serine/arginine repetitive matrix protein 4 (UniProt: A7MD48; also known as Medulloblastoma antigen MU-MB-2.76, Neural-specific serine/arginine repetitive splicing factor of 100 kDa, Neural-specific SR-related protein of 100 kDa, nSR100) is encoded by the SRRM4 (also known as KIAA1853) gene (Gene ID: 84530) in human. nSR100 is a nuclear protein that is specifically expressed in neuronal cells. It serves as a splicing factor that is specifically required for neural cell differentiation. It acts in conjunction with nPTB/PTBP2 by binding directly to its regulated target transcripts and promote neural-specific exon inclusion in many genes that function in neural cell differentiation. It is shown to regulate the splicing of protrudin gene (Zfyve27) transcripts in neuronal cells and promotes splicing of protrudin pre-mRNA so as to include a microexon (exon L) encoding seven amino acids in a neuron-specific manner. The resulting protein known as Protrudin-L promotes neurite outgrowth during neurogenesis. Depletion of nSR100 in Neuro2A cells is shown to impair neurite outgrowth, whereas its ablation in mouse embryos is reported to prevent differentiation of neuronal progenitors in the cortex and impair development of the central and peripheral nervous systems. (Ref.: Ohnishi, T et al. (2017). Sci. Rep. 7: 41130; Quesnel-Vallières, M et al. (2015). Genes Dev. 29(7); 746-759).
Specifications
nSR100 (SRRM4) | |
Polyclonal | |
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected nSR100 (SRRM4) in mouse cerebral cortex and human cerebral cortex tissue sections. | |
Serine/arginine repetitive matrix protein 4;Medulloblastoma antigen MU-MB-2.76;Neural-specific serine/arginine repetitive splicing factor of 100 kDa;Neural-specific SR-related protein of 100 kDa;nSR100 | |
Rabbit | |
∽68 kDa observed; 68.56 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Neuroscience | |
This rabbit polyclonal antibody detects Serine/arginine repetitive matrix protein 4 in human and mouse cells. It targets an epitope within 104 amino acids from the C-terminal half. | |
Stable for 1 year at 2-8°C from date of receipt. |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
SRRM4;KIAA1853 | |
GST/His-tagged recombinant frgment corresponding to 104 amino acids from the C-terminal half of human Serine/arginine repetitive matrix protein 4. | |
Affinity Purified | |
RUO | |
Primary | |
Mouse, Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction