Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSrp70 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568315
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8315 Supplier Bethyl Laboratories, Inc Supplier No. A304658A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

RNA-binding protein that mediates pre-mRNA alternative splicing regulation.

Specifications

Antigen NSrp70
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NSRP1
Gene Accession No. Q9H0G5
Gene Alias Nuclear speckle splicing regulatory protein 1, Coiled-coil domain-containing protein 55, Nuclear speckle-related protein 70, NSrp70, NSRP1, CCDC55, NSRP70
Gene Symbols NSRP1
Host Species Rabbit
Immunogen Between 508 and 558
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84081
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.