Learn More
Invitrogen™ NSUN2 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
Specifications
Specifications
| Antigen | NSUN2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | NSUN2 |
| Gene Accession No. | Q08J23 |
| Gene Alias | 5-methycytoisine methyltransferase; D13Wsu123e; hTrm4; Misu; mRNA cytosine C(5)-methyltransferase; MRT5; Myc-induced SUN domain containing protein; myc-induced SUN domain-containing protein; Myc-induced SUN-domain-containing protein; NOL1/NOP2/Sun domain family 2; NOL1/NOP2/Sun domain family member 2; NOL1/NOP2/Sun domain family, member 2; NOP2/Sun domain family, member 2; NOP2/Sun RNA methyltransferase family member 2; NOP2/Sun RNA methyltransferase family, member 2; Nsun2; RGD1311954; RNA cytosine C(5)-methyltransferase NSUN2; SAKI; substrate of AIM1/Aurora kinase B; TRM4; tRNA (cytosine(34)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase NSUN2; tRNA cytosine C(5)-methyltransferase; tRNA methyltransferase 4 homolog |
| Gene Symbols | NSUN2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.