Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSUN3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NSUN3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NSUN3 Polyclonal specifically detects NSUN3 in Human samples. It is validated for Western Blot.Specifications
NSUN3 | |
Polyclonal | |
Rabbit | |
Q9H649 | |
63899 | |
Synthetic peptides corresponding to NSUN3(NOL1/NOP2/Sun domain family, member 3) The peptide sequence was selected from the middle region of NSUN3. Peptide sequence GGKSIALLQCACPGYLHCNEYDSLRLRWLRQTLESFIPQPLINVIKVSEL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.1.1, EC 2.1.1.-, FLJ22109, FLJ22609, MST077, NOL1/NOP2/Sun domain family 3, NOL1/NOP2/Sun domain family member 3, NOL1/NOP2/Sun domain family, member 3, NOP2/Sun domain family, member 3, putative methyltransferase NSUN3 | |
NSUN3 | |
IgG | |
38 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title