Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSUN4 Monoclonal Antibody (4G4)
GREENER_CHOICE

Catalog No. PIMA542532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542532 Supplier Invitrogen™ Supplier No. MA542532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: MCF-7 cell lysate, 293 cell lysate, mouse testis tissue, human prostate tissue. Subcellular Location: Mitochondrion.

NSUN4 is a 5-methylcytosine rRNA methytransferase that is involved in mitochondiral ribosome assembly. More specifically NSUN4 is involved in mitochondrial ribosome small subunit (SSU) maturation by methylation of mitochondrial 12S rRNA. This function is independent of MTREFD2/MTERF4 and assembled mitochondrial ribosome large subunit (LSU). NSUN4 is targeted to LSU by MTREFD2/MTERF4 and is involved in the final step in ribosome biogenesis to ensure that SSU and LSU are assembled.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSUN4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4G4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene NSUN4
Gene Accession No. Q96CB9, Q9CZ57
Gene Alias 2310010O12Rik; 2810405F18Rik; 5-methylcytosine rRNA methyltransferase NSUN4; AI507900; NOL1/NOP2/Sun domain family member 4; NOL1/NOP2/Sun domain family, member 4; NOP2/Sun domain family, member 4; NOP2/Sun RNA methyltransferase family member 4; Nsun4; putative methyltransferase NSUN4; RGD1559652; RP4-603I14.2; SHTAP; Sperm head and tail associated protein
Gene Symbols NSUN4
Host Species Mouse
Immunogen Recombinant protein within human NSUN4 aa 1-384.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387338, 72181
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.