Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSUN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546042 Supplier Invitrogen™ Supplier No. PA546042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ASPETTLSSG FFVAVIERVE VPSSASQAKA SAPERTPSPA PKRKKRQQRA Sequence homology: Human: 100%.

NSUN5 is a member of the evolutionarily conserved NOL1/NOP2/Sun domain family. The encoded protein may function as a DNA methyltransferase in the nucleus. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSUN5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nsun5
Gene Accession No. Q96P11
Gene Alias 9830109N13Rik; AI326939; FLJ10267; MGC986; NOL1; NOL1/NOP2/Sun domain family member 5; NOL1/NOP2/Sun domain family, member 5; NOL1R; NOL1-related protein; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5A; NOP2/Sun RNA methyltransferase family member 5; Nsun5; NSUN5A; p120; p120(NOL1); Probable 28S rRNA (cytosine-C(5))-methyltransferase; putative methyltransferase NSUN5; RGD1309268; WBSCR20; Wbscr20a; Williams Beuren syndrome chromosome region 20A; Williams Beuren syndrome chromosome region 20A homolog; Williams-Beuren syndrome chromosomal region 20A protein; Williams-Beuren syndrome chromosomal region 20A protein homolog; Williams-Beuren syndrome critical region protein 20; Williams-Beuren syndrome critical region protein 20 copy A
Gene Symbols Nsun5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human NSUN5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55695
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.