Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSUN5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 154491AP150UL

Description
NSUN5 is a member of the evolutionarily conserved NOL1/NOP2/Sun domain family. The encoded protein may function as a DNA methyltransferase in the nucleus. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.Specifications
NSUN5 | |
Polyclonal | |
Unconjugated | |
NSUN5 | |
NOL1, NOL1 related protein, NOL1R, NSUN5, NSUN5A, p120, WBSCR20, WBSCR20A, Ynl022cL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100609, 288595, 55695 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4F6, Q96P11 | |
Nsun5 | |
NSUN5 Fusion Protein Ag7711 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction