Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSUN5 Recombinant Rabbit Monoclonal Antibody (9H9L15)

Catalog No. PI702063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI702063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI702063 Supplier Thermo Scientific Supplier No. 702063
Add to Cart
Add to Cart

NSUN5 Recombinant Rabbit Monoclonal Antibody (9H9L15)

NSUN5 Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NSUN5
Applications Western Blot
Classification Recombinant Monoclonal
Clone 9H9L15
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Nsun5
Gene Accession No. Q96P11
Gene Alias 9830109N13Rik; AI326939; FLJ10267; MGC986; NOL1; NOL1/NOP2/Sun domain family member 5; NOL1/NOP2/Sun domain family, member 5; NOL1R; NOL1-related protein; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5A; NOP2/Sun RNA methyltransferase family member 5; Nsun5; NSUN5A; p120; p120(NOL1); Probable 28S rRNA (cytosine-C(5))-methyltransferase; putative methyltransferase NSUN5; RGD1309268; WBSCR20; Wbscr20a; Williams Beuren syndrome chromosome region 20A; Williams Beuren syndrome chromosome region 20A homolog; Williams-Beuren syndrome chromosomal region 20A protein; Williams-Beuren syndrome chromosomal region 20A protein homolog; Williams-Beuren syndrome critical region protein 20; Williams-Beuren syndrome critical region protein 20 copy A
Gene Symbols Nsun5
Host Species Rabbit
Immunogen Protein corresponding to human NSUN5 (aa150-aa429).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55695
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.