Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSUN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561119 Supplier Invitrogen™ Supplier No. PA561119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLLIPVIGPR KNIKKQQCEA IVGAQCGNAV LRGAHVYAPG IVSASQFMKA GDVISVYSDI KGKCKKGAKE FDGTKVFLGN GISELSRKEI FSG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

NSun6 is an RNA methyltransferase that catalyzes the methylation at position cytosine C72 in some tRNAs. NSUN6 has been reported to localize to the cytoplasm and largely colocalize with Golgi apparatus and pericentriolar matrix, indicating that NSUN6 modifies tRNAs in a late step in their biogenesis. NSun6 may also play a major role in bone metastasis and could be a valuable therapeutic target for bone metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSUN6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NSUN6
Gene Accession No. Q8TEA1
Gene Alias 4933403D21Rik; 4933414E04Rik; ARL5B antisense RNA 1; ARL5B-AS1; NOL1/NOP2/Sun and PUA domain-containing protein 1; NOL1/NOP2/Sun domain family 6; NOL1/NOP2/Sun domain family member 6; NOL1/NOP2/Sun domain family, member 6; NOP2/Sun domain family, member 6; NOP2/Sun RNA methyltransferase family member 6; NOPD1; NSUN6; nucleolar protein (NOL1/NOP2/sun) and PUA domains 1; putative methyltransferase NSUN6
Gene Symbols NSUN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NSUN6. Recombinant protein control fragment (Product #RP-97123).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221078
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.