Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT-3 Antibody (OTI5A2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272972F
Description
NT-3 Monoclonal specifically detects NT-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| NT-3 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NTF3 | |
| Recombinant protein expressed in E.coli corresponding to amino acids 139-257 of human Neurotrophin 3. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5A2 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 4908.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction