Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT-3 Antibody (OTI5A2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147892
Description
NT-3 Monoclonal specifically detects NT-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Neurotrophin 3 | |
Monoclonal | |
1.5 mg/mL | |
Western Blot 1:5000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
NTF3 | |
Recombinant protein expressed in E.coli corresponding to amino acids 139-257 of human NT3 | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens neurotrophin 3 (NTF3), transcript variant 1. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI5A2 | |
Unconjugated | |
HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3 | |
Mouse | |
27.4 kDa | |
0.1 mL | |
Apoptosis, Cancer | |
4908 | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction