Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT-proBNP Antibody (13G12cc), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12014712
Description
NT-proBNP Monoclonal specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
NT-proBNP | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4) with 0.09% Sodium Azide | |
NPPB | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 13-27, conjugated with carrier protein. | |
0.2 mg | |
Cytokine Research, Immunology, Neuroscience | |
4879 | |
Human | |
Purified |
Western Blot, ELISA | |
13G12cc | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human NT-proBNP and proBNP | |
Store at 4C. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction