Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT-proBNP Antibody (OTI11G4), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271213V
Description
NT-proBNP Monoclonal specifically detects NT-proBNP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| NT-proBNP | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NPPB | |
| Recombinant protein fragment corresponding to amino acids 27-102 of human BNP (NP_002512) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI11G4 | |
| Western Blot, Immunohistochemistry | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
| 4879.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction