Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT3 Mouse anti-Human, Clone: OTI5A2, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500030

Description
The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintece of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.Specifications
NT3 | |
Monoclonal | |
1.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P20783 | |
NTF3 | |
Recombit protein expressed in E.coli corresponding to amino acids 139-257 of human NT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5A2 | |
Unconjugated | |
NTF3 | |
HDNF, NGF-2, NGF2, NT-3, NT3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4908 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction