Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5C1A Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306431JF549
This item is not returnable.
View return policy
Description
NT5C1A Polyclonal antibody specifically detects NT5C1A in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NT5C1A | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
5'-nucleotidase, cytosolic IA, AMP-specific 5'-NT, CN1, cN1A, CN-I, cN-IA, cytosolic 5' nucleotidase, type 1A, cytosolic 5'-nucleotidase 1A, Cytosolic 5'-nucleotidase IA, EC 3.1.3.5, MGC119199, MGC119201 | |
Produced in rabbits immunized with E. coli-derived Human NT5C1A fragment. | |
0.1 mL | |
Lipid and Metabolism | |
84618 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction