Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5C1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562163 Supplier Invitrogen™ Supplier No. PA562163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVWEEAKIFY DNLAPKKKPK SPKPQNAVTI AVSSRALFRM DEEQQIYTEQ GVEEYVRYQL EHENEPFSPG P Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

Dephosphorylates the 5' and 2'(3')-phosphates of deoxyribonucleotides and has a broad substrate specificity. Helps to regulate adenosine levels in heart during ischemia and hypoxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5C1A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NT5C1A
Gene Accession No. Q9BXI3
Gene Alias 5'-nucleotidase, cytosolic IA; AMP-specific 5'-NT; CN1; CN1A; CNI; CN-I; cN-IA; cytosolic 5' nucleotidase, type 1A; cytosolic 5'-nucleotidase 1A; Cytosolic 5'-nucleotidase IA; Gm49; NT5C1A
Gene Symbols NT5C1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NT5C1A. Recombinant protein control fragment (Product #RP-101219).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.