Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5C1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563340 Supplier Invitrogen™ Supplier No. PA563340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQTSLKQKKN EPGMRSSKES LEAEKRKESD KTGVRLSNQM RRAVNPNHSL RCCPFQGHSS CRRCLCAAEG TALGPCHTIR IYIHMCLLWE QG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.

Cytosolic 5-prime nucleotidases, such as NT5C1B, catalyze production of adenosine, which regulates diverse physiologic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5C1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NT5C1B
Gene Accession No. Q96P26
Gene Alias 4921514H13Rik; 5'nucleotidase; 5'-nucleotidase, cytosolic IB; AIRP; Autoimmune infertility-related protein; CN1B; cN-IB; cytosolic 5'-nucleotidase 1B; Cytosolic 5'-nucleotidase IB; FKSG85; NT5C1B; testicular tissue protein Li 1
Gene Symbols NT5C1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NT5C1B. Recombinant protein control fragment (Product #RP-100561).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.