Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NT5C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530456 Supplier Invitrogen™ Supplier No. PA530456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, HCT116. Predicted reactivity: Mouse (100%), Xenopus laevis (91%), Chicken (97%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Purine 5-prime-nucleotidase (EC 3.1.3.5) preferentially hydrolyzes inosine 5-prime-monophosphate (IMP) and other purine nucleotides, and is allosterically activated by various compounds, including ATP. The enzyme is exclusively located in the cytoplasmic matrix of cells and may have a critical role in the maintenance of a constant composition of intracellular purine/pyrimidine nucleotides in cooperation with other nucleotidases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5C2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene NT5C2
Gene Accession No. P49902
Gene Alias 2010002I23Rik; 5'-nucleotidase (purine), cytosolic type B; 5'-nucleotidase, cytosolic II; CnII; cN-II; Cytosolic 5'-nucleotidase II; cytosolic purine 5'-nucleotidase; ENSMUSG00000025039; Gm9751; Gmp; IMP-specific 5'-NT; Nt5b; Nt5c2; NT5CP; PNT5; RP11-30H12.3; SPG45; SPG65
Gene Symbols NT5C2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human NT5C2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.