Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NT5C2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112865
Description
NT5C2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Purine 5-prime-nucleotidase (EC 3.1.3.5) preferentially hydrolyzes inosine 5-prime-monophosphate (IMP) and other purine nucleotides, and is allosterically activated by various compounds, including ATP. The enzyme is exclusively located in the cytoplasmic matrix of cells and may have a critical role in the maintenance of a constant composition of intracellular purine/pyrimidine nucleotides in cooperation with other nucleotidases.
Specifications
| NT5C2 | |
| Polyclonal | |
| Unconjugated | |
| NT5C2 | |
| 2010002I23Rik; 5'-nucleotidase (purine), cytosolic type B; 5'-nucleotidase, cytosolic II; CnII; cN-II; Cytosolic 5'-nucleotidase II; cytosolic purine 5'-nucleotidase; ENSMUSG00000025039; Gm9751; Gmp; IMP-specific 5'-NT; Nt5b; Nt5c2; NT5CP; PNT5; RP11-30H12.3; SPG45; SPG65 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 22978, 76952 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.77 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P49902, Q3V1L4 | |
| NT5C2 | |
| Recombinant Human Cytosolic purine 5'-nucleotidase protein (487-553AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction