Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5C2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980235
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0235 Supplier Bioss Supplier No. BS0372R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purine 5-prime-nucleotidase (EC 3.1.3.5) preferentially hydrolyzes inosine 5-prime-monophosphate (IMP) and other purine nucleotides, and is allosterically activated by various compounds, including ATP. The enzyme is exclusively located in the cytoplasmic matrix of cells and may have a critical role in the maintenance of a constant composition of intracellular purine/pyrimidine nucleotides in cooperation with other nucleotidases.

Specifications

Antigen NT5C2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NT5C2
Gene Accession No. P49902
Gene Alias 2010002I23Rik; 5′-nucleotidase (purine), cytosolic type B; 5′-nucleotidase, cytosolic II; CnII; cN-II; Cytosolic 5′-nucleotidase II; Cytosolic purine 5′-nucleotidase; ENSMUSG00000025039; Gm9751; Gmp; IMP-specific 5′-NT; NT5B; NT5C2; NT5CP; PNT5; RP11-30H12.3; SPG45; SPG65
Gene Symbols NT5C2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NT5C2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22978
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.